Learn More
Cayman Chemical Thiodigalactoside, Purity: ≥98%, CAS: 51555-87-4, 100 mg
SDP
Catalog No. 502966590
Encompass_Preferred
A diglycan that binds galectins-1, -3, -8, and -9 with Kd values between 24 and 78 µM; used to investigate the roles of these galectins in cell signaling and carcinogenesis
Catalog No. 50-296-6590 Supplier Cayman Chemical Supplier No. 17154100 MG
A diglycan that binds galectins-1, -3, -8, and -9 with Kd values between 24 and 78 µM; used to investigate the roles of these galectins in cell signaling and carcinogenesis