Learn More
Cayman Chemical Thiodigalactoside, Purity: ≥98%, CAS: 51555-87-4, 50 mg
SDP
Catalog No. 502979505
Encompass_Preferred
A diglycan that binds galectins-1, -3, -8, and -9 with Kd values between 24 and 78 µM; used to investigate the roles of these galectins in cell signaling and carcinogenesis
Catalog No. 50-297-9505 Supplier Cayman Chemical Supplier No. 1715450 MG
A diglycan that binds galectins-1, -3, -8, and -9 with Kd values between 24 and 78 µM; used to investigate the roles of these galectins in cell signaling and carcinogenesis