Learn More
Cayman Chemical Thiodiglycolic Acid-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 132090-51-8, 5 mg
SDP
Catalog No. 502894385
Encompass_Preferred
An internal standard for the quantification of TDGA by GC- or LC-MS
Catalog No. 50-289-4385 Supplier Cayman Chemical Supplier No. 90024785 MG
An internal standard for the quantification of TDGA by GC- or LC-MS