Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiodisuccinic Acid 98.0+%, TCI America™
SDP

Catalog No. T089725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T089725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T089725G Supplier TCI America Supplier No. T089725G

Chemical Identifiers

CAS 4917-76-4
Molecular Formula C8H10O8S
Molecular Weight (g/mol) 266.22
MDL Number MFCD00020502
InChI Key SASYRHXVHLPMQD-UHFFFAOYNA-N
Synonym thiodisuccinic acid, 2,2'-thiodisuccinic acid, 2,2-thiodisuccinic acid, butanedioic acid, 2,2'-thiobis, 2-1,2-dicarboxyethyl sulfanyl butanedioic acid, succinic acid, 2,2'-thiodi, 2,2'-sulfanediyldisuccinic acid, thiodisuccinicacid, pubchem15325, acmc-1ao15
PubChem CID 107352
IUPAC Name 2-[(1,2-dicarboxyethyl)sulfanyl]butanedioic acid
SMILES OC(=O)CC(SC(CC(O)=O)C(O)=O)C(O)=O

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 266.22
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Thiodisuccinic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.