Learn More
Cayman Chemical ThIoetheramIde-PC 10mg
SDP
Catalog No. 502896435
Encompass_Preferred
Thioetheramide-PC is a structurally modified phospholipid that functions as a competitive, reversible inhibitor of sPLA2.{808,2624} The IC50 value for thioetheramide-PC is 2 µM at a substrate concentration of 0.5 mM.{808} In addition to binding to the catalytic site of sPLA2, thioetheramide-PC also binds to the activator site of this enzyme. The binding of thioetheramide-PC to the activator site is tighter than its binding to the catalytic site. The result of this dual interaction is that at low concentrations thioetheramide-PC may activate phospholipase activity rather than inhibiting it.{808}
Catalog No. 50-289-6435 Supplier Cayman Chemical Supplier No. 6275010 MG
Thioetheramide-PC is a structurally modified phospholipid that functions as a competitive, reversible inhibitor of sPLA2.{808,2624} The IC50 value for thioetheramide-PC is 2 µM at a substrate concentration of 0.5 mM.{808} In addition to binding to the catalytic site of sPLA2, thioetheramide-PC also binds to the activator site of this enzyme. The binding of thioetheramide-PC to the activator site is tighter than its binding to the catalytic site. The result of this dual interaction is that at low concentrations thioetheramide-PC may activate phospholipase activity rather than inhibiting it.{808}