Learn More
Enzo Life Sciences Thioetheramide-phosphorylcholine (5 mg). CAS: 116457-99-9
SDP
Supplier:  Enzo Life Sciences BMLST3250005
Encompass_Preferred
A potent substrate analog inhibitor of snake venom PLA2 (IC50=2 µM). It inhibits human recombinant sPLA2 (IC50=0.9 µM) selectively over cPLA2. Alternative name: 1-Palmitylthio-2-palmitoylamido-1,2-dideoxy-sn-glycero-3-phosphorylcholine. Purity: ≥98% (TLC). Solubility: Soluble in DMSO, 100% ethanol or methanol. Long Term Storage: -20°C.
Catalog No. 50-201-2158