Learn More
Cayman Chemical ThIoflavIn T 10g
SDP
Catalog No. 502951037
Encompass_Preferred
A fluorescent probe for amyloid fibrils; ex/em = 385/445 nm, respectively, which increases to 450/482 upon amyloid fibril binding with an increase in intensity; intensity is pH dependent, with increasing intensity as the pH increases; binding affinity varies with fibril type and binding mode; has been used to visualize amyloid fibrils in rat brain in a model of Alzheimer's disease
Catalog No. 50-295-1037 Supplier Cayman Chemical Supplier No. 3255310 G
A fluorescent probe for amyloid fibrils; ex/em = 385/445 nm, respectively, which increases to 450/482 upon amyloid fibril binding with an increase in intensity; intensity is pH dependent, with increasing intensity as the pH increases; binding affinity varies with fibril type and binding mode; has been used to visualize amyloid fibrils in rat brain in a model of Alzheimer's disease