Learn More
Medchemexpress LLC Thioflavin T | 2390-54-7 | 99.4% | 500 MG
SDP
Catalog No. 5000409858
Encompass_Preferred
Thioflavin T is a cationic Benzothiazole dye that exhibits enhanced fluorescence upon binding to amyloid in tissue sections. It is a fluorescent dye used for staining Aβ fibrils with green fluorescence and is commonly used to measure protein fibrillation. Weakly fluorescent in aqueous solutions, it shows enhanced emission at 482 nm when bound to Aβ plaques. It can be applied in Alzheimer's disease research and to detect endoplasmic reticulum stress (ERS) by displaying green fluorescence when binding to protein aggregates.
  • Cationic Benzothiazole dye
  • Enhanced fluorescence upon binding to amyloid in tissue sections
  • Excitation max. approximately 385 nm (free) and 450 nm (bound)
  • Emission max. approximately 445 nm (free) and 485 nm (bound)
  • Stains Aβ fibrils with green fluorescence
  • Detects endoplasmic reticulum stress (ERS)
  • Useful for measuring protein fibrillation
Catalog No. 50-004-09858 Supplier Medchemexpress LLC Supplier No. HYD0218500MG
Thioflavin T is a cationic Benzothiazole dye that exhibits enhanced fluorescence upon binding to amyloid in tissue sections. It is a fluorescent dye used for staining Aβ fibrils with green fluorescence and is commonly used to measure protein fibrillation. Weakly fluorescent in aqueous solutions, it shows enhanced emission at 482 nm when bound to Aβ plaques. It can be applied in Alzheimer's disease research and to detect endoplasmic reticulum stress (ERS) by displaying green fluorescence when binding to protein aggregates.
  • Cationic Benzothiazole dye
  • Enhanced fluorescence upon binding to amyloid in tissue sections
  • Excitation max. approximately 385 nm (free) and 450 nm (bound)
  • Emission max. approximately 445 nm (free) and 485 nm (bound)
  • Stains Aβ fibrils with green fluorescence
  • Detects endoplasmic reticulum stress (ERS)
  • Useful for measuring protein fibrillation