Learn More
Abcam Thioflavin T, Fluorescent cell-permeable amyloid binding benzothiazole salt, 500MG
Catalog No. NC0517036
Encompass_Preferred
MW 318.9 Da, Purity >97%. Fluorescent cell-permeable amyloid binding benzothiazole salt. Dye used to visualise stacked β sheets in vitro and in vivo. Binding induces a shift in excitation maximum (385 nm to 450 nm) and emission maximum (445 nm to 482 nm). Highly soluble in water. Limited ability to cross the blood-brain barrier.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0517036 Supplier Abcam Supplier No. AB120751500MG
MW 318.9 Da, Purity >97%. Fluorescent cell-permeable amyloid binding benzothiazole salt. Dye used to visualise stacked β sheets in vitro and in vivo. Binding induces a shift in excitation maximum (385 nm to 450 nm) and emission maximum (445 nm to 482 nm). Highly soluble in water. Limited ability to cross the blood-brain barrier.
The product is subject to the following: Abcam Restricted Use Statement