Learn More
Biotium Thioflavin T "high purity grade"
Catalog No. 501964787
Encompass_Preferred
A cell-permeable benzothiazole dye that exhibits enhanced fluorescence (Ex/Em 450/482 nm) upon binding to amyloid fibrils. Useful in monitoring stacked b-sheet aggregates, Thioflavin T has also been used in histology and for protein characterization.Biotium offers high purity grade of Thioflavin T with HPLC purity greater than 97%.Properties: Yellow solid soluble in DMF and DMSO; Stored desiccated at 4°C; C17H19ClN2S; MWt: 318.86; [2390-54-7]
Catalog No. 50-196-4787 Supplier Biotium Supplier No. 80033
A cell-permeable benzothiazole dye that exhibits enhanced fluorescence (Ex/Em 450/482 nm) upon binding to amyloid fibrils. Useful in monitoring stacked b-sheet aggregates, Thioflavin T has also been used in histology and for protein characterization.Biotium offers high purity grade of Thioflavin T with HPLC purity greater than 97%.Properties: Yellow solid soluble in DMF and DMSO; Stored desiccated at 4°C; C17H19ClN2S; MWt: 318.86; [2390-54-7]