Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thioflavin T, Sigma-Aldrich

Catalog No. 01259065 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-259-065 5 g
01-259-042 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-259-065 Supplier MilliporeSigma Supelco Supplier No. T35165G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thioflavin T is a benzothiazole dye that increases in fluorescence upon binding to amyloid. Thioflavin T has been used in histology and for protein characterization.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2390-54-7
Molecular Weight (g/mol) 318.86 g/mol

Specifications

Color Brown to Yellow
Quantity 5 g
Chemical Name or Material Thioflavin T
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.