Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thioflavin T UV-VIS Internal Standard in Water, 18 ppm, SPEX CertiPrep™
SDP

Catalog No. UV413W Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
3.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
UV413W 3.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. UV413W Supplier SPEX CertiPrep Supplier No. UV413W

missing translation for 'validMainframeMsg'

UV-VIS internal standards allow the user to accurately measure the concentration of their samples by directly comparing the absorbance of their compounds against that of the internal standard.

SPEX Certiprep™ offers eight different standards that cover a wide range of wavelengths (270 to 627 nm). After the user takes a UV-VIS spectrum of their sample, they can find a region in the spectrum that doesn’t absorb, and then select the corresponding internal standard, and rerun the spectrum.

Specifications

Chemical Name or Material Thioflavin T
Packaging Ampoule
CAS 2390-54-7
Molecular Formula C17H19ClN2S
Concentration 18 ppm
Technique UV/Vis, Spectrometry, LCMS and HPLC
Color Colorless to Light Yellow
Synonym UV-413 nm internal standard
Solubility Soluble in water
Type UV Internal Standard
Molecular Weight (g/mol) 318.9
Quantity 3.5 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.