Learn More
Thioflavine S, Practical Grade, Sigma-Aldrich
Catalog No. 01259066 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
Catalog No. Quantity
01-259-066 25 g
1 options
Catalog No. 01-259-066 Supplier MilliporeSigma Supelco Supplier No. T189225G
Add to Cart
Add to Cart
Stain for visualizing β-amyloid structures in tissue sections.
Thioflavine S aids in staining and visualizing B-amyloid plaques in Alzheimer's disease. Thioflavin S binds to fibrillar B-amyloid plaques, and does not diffuse in the plaques. Thioflavin-S staining aids in visualizing smaller plaques throughout the neocortex, hippocampus and thalamic region.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 25 g
Product Type Stain Kit