Learn More
Medchemexpress LLC Thioglycolate sodiu 10g
SDP
Catalog No. 5000206741
Encompass_Preferred
Thioglycolate sodium is used in inflammation research to elicit a neutrophil and macrophage response in vivo Thioglycolate sodium can induce peritonitis in vivo[1][2]
Catalog No. 50-002-06741 Supplier Medchemexpress LLC Supplier No. HYW11572410G
Thioglycolate sodium is used in inflammation research to elicit a neutrophil and macrophage response in vivo Thioglycolate sodium can induce peritonitis in vivo[1][2]