Learn More
Medchemexpress LLC Thioglycolate sodiu 25g
SDP
Catalog No. 5000204867
Encompass_Preferred
Thioglycolate sodium is used in inflammation research to elicit a neutrophil and macrophage response in vivo Thioglycolate sodium can induce peritonitis in vivo[1][2]
Catalog No. 50-002-04867 Supplier Medchemexpress LLC Supplier No. 25GHYW115724
Thioglycolate sodium is used in inflammation research to elicit a neutrophil and macrophage response in vivo Thioglycolate sodium can induce peritonitis in vivo[1][2]