Learn More
Medchemexpress LLC Thioglycolate sodiu 5g
SDP
Catalog No. 5000206479
Encompass_Preferred
Thioglycolate sodium is used in inflammation research to elicit a neutrophil and macrophage response in vivo Thioglycolate sodium can induce peritonitis in vivo[1][2]
Catalog No. 50-002-06479 Supplier Medchemexpress LLC Supplier No. HYW1157245G
Thioglycolate sodium is used in inflammation research to elicit a neutrophil and macrophage response in vivo Thioglycolate sodium can induce peritonitis in vivo[1][2]