Learn More
Alkali Scientific Thioglycollate w/o Indole w/Hemin&Vitamin K 10mL Tube, 10/Pk
SDP
Supplier:  Alkali Scientific TWOT00
Encompass_Preferred
Thioglycollate medium enriched with hemin and vitamin K is a non selective liquid medium designed for cultivating fastidious anaerobic bacteria from clinical samples. The added growth factors support organisms that require specific nutrients for survival and growth. The formulation creates an oxygen reduced environment suitable for anaerobic organisms while maintaining conditions that allow recovery of a broad range of bacteria. This medium is commonly used in clinical laboratories for processing tissue samples, wound specimens, and other materials where anaerobic pathogens may be present. It supports reliable recovery and identification of clinically significant microorganisms.
Catalog No. 50-006-62472