Learn More
Arbor Assays Thiol Fluorescent Detection Kit (1 Plate)
SDP
Catalog No. 502278382
Encompass_Preferred
The DetectX Thiol Fluorescent Detection Kit allows users to accurately determine the extent of free thiol content in samples using a proprietary substrate, ThioStar, that is converted to a brightly fluorescent product upon reaction with thiols in the sample.
This item is not returnable. View return policy
Catalog No. 50-227-8382 Supplier Arbor Assays Supplier No. K005F1
The DetectX Thiol Fluorescent Detection Kit allows users to accurately determine the extent of free thiol content in samples using a proprietary substrate, ThioStar, that is converted to a brightly fluorescent product upon reaction with thiols in the sample.