Learn More
Medchemexpress LLC Thiol-PEG-acetic acid (SH-PEG-COOH) | 00-00-0 | 95.0% | HSC2H4O(C2H4O)nCH2CO2H | 10MG
SDP
Catalog No. 5000260437
Encompass_Preferred
Thiol-PEG-CH2COOH (MW 5000) is a heterobifunctional polyethylene glycol linker bearing a thiol at one terminus and a carboxylic acid at the other. It provides a flexible spacer for PROTAC assembly and other bioconjugation workflows, enabling orthogonal attachment strategies while maintaining solubility and spacer length control.
Catalog No. 50-002-60437 Supplier Medchemexpress LLC Supplier No. HY14073010MG
Thiol-PEG-CH2COOH (MW 5000) is a heterobifunctional polyethylene glycol linker bearing a thiol at one terminus and a carboxylic acid at the other. It provides a flexible spacer for PROTAC assembly and other bioconjugation workflows, enabling orthogonal attachment strategies while maintaining solubility and spacer length control.