Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thiol-PEG-CH2COOH | 95.0% | HS-(CH2-CH2-O)n-CH2COOH | 25 MG
SDP

Catalog No. 5000352515
Encompass_Preferred

Thiol-PEG-CH2COOH (MW 5000) is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. PROTACs utilize two different ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein, to exploit the intracellular ubiquitin-proteasome system for selective degradation of target proteins.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits intracellular ubiquitin-proteasome system
  • Target proteins selectively degraded
  • Average molecular weight of 5000
  • Purity of 95.0% by NMR
  • Appearance is white to off-white solid

Catalog No. 50-003-52515 Supplier Medchemexpress LLC Supplier No. 25MGHY140730
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thiol-PEG-CH2COOH (MW 5000) is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. PROTACs utilize two different ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein, to exploit the intracellular ubiquitin-proteasome system for selective degradation of target proteins.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits intracellular ubiquitin-proteasome system
  • Target proteins selectively degraded
  • Average molecular weight of 5000
  • Purity of 95.0% by NMR
  • Appearance is white to off-white solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.