Learn More
Medchemexpress LLC Thiol-PEG-CH2COOH (MW 2000) | 95.0% | 100 MG
SDP
Catalog No. 5000355494
Encompass_Preferred
Thiol-PEG-CH2COOH (MW 2000) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-55494 Supplier Medchemexpress LLC Supplier No. HY140728100MG
Add to Cart
Add to Cart
Thiol-PEG-CH2COOH (MW 2000) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins