Learn More
Medchemexpress LLC Thiol-PEG-CH2COOH (MW 2000) | 95.0% | 25 MG
SDP
Catalog No. 5000352624
Encompass_Preferred
Thiol-PEG-CH2COOH (MW 2000) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs.
  • PROTACs contain two different ligands connected by a linker.
  • One ligand is for an E3 ubiquitin ligase and the other is for the target protein.
  • PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
Catalog No. 50-003-52624 Supplier Medchemexpress LLC Supplier No. 25MGHY140728
Thiol-PEG-CH2COOH (MW 2000) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs.
  • PROTACs contain two different ligands connected by a linker.
  • One ligand is for an E3 ubiquitin ligase and the other is for the target protein.
  • PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.