Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thiol-PEG-CH2COOH (MW 2000) | 95.0% | 50 MG
SDP

Catalog No. 5000352960
Encompass_Preferred

Thiol-PEG-CH2COOH (MW 2000) is a PEG-based PROTAC linker. It features a thiol group at one end and a carboxymethyl group at the other, making it suitable for crosslinking and PROTAC synthesis. This compound is used in laboratory research for various applications requiring specific chemical modifications and conjugation.

  • PROTAC linker
  • Thiol-functionalized
  • Carboxymethyl-functionalized
  • Used in synthesis of PROTACs
  • Molecular weight of 2000
  • Purity of 95.0%
  • Storage as powder at -20°C for 3 years
  • Storage in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-003-52960 Supplier Medchemexpress LLC Supplier No. HY14072850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thiol-PEG-CH2COOH (MW 2000) is a PEG-based PROTAC linker. It features a thiol group at one end and a carboxymethyl group at the other, making it suitable for crosslinking and PROTAC synthesis. This compound is used in laboratory research for various applications requiring specific chemical modifications and conjugation.

  • PROTAC linker
  • Thiol-functionalized
  • Carboxymethyl-functionalized
  • Used in synthesis of PROTACs
  • Molecular weight of 2000
  • Purity of 95.0%
  • Storage as powder at -20°C for 3 years
  • Storage in solvent at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.