Learn More
Medchemexpress LLC Thiol-PEG-CH2COOH (MW 5000) | 95.0% | SMILES: SCCOCC(O)=O.[MW 5000].[n] | 50 MG
SDP
Catalog No. 5000353518
Encompass_Preferred
Thiol-PEG-CH2COOH (MW 5000) is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are composed of two distinct ligands connected by a linker, one targeting an E3 ubiquitin ligase and the other targeting the protein intended for degradation, leveraging the intracellular ubiquitin-proteasome system for selective protein degradation.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Leverages the intracellular ubiquitin-proteasome system
  • Enables selective degradation of target proteins
Catalog No. 50-003-53518 Supplier Medchemexpress LLC Supplier No. HY14073050MG
Thiol-PEG-CH2COOH (MW 5000) is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are composed of two distinct ligands connected by a linker, one targeting an E3 ubiquitin ligase and the other targeting the protein intended for degradation, leveraging the intracellular ubiquitin-proteasome system for selective protein degradation.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Leverages the intracellular ubiquitin-proteasome system
  • Enables selective degradation of target proteins