Learn More
LAYSAN BIO INC Thiol-PEG-Thiol, 3400 Da, 5 g, Linear, Homobifunctional, Powder, Highly soluble in water
Catalog No. NC2455867
Encompass
LAYSAN BIO INC's Thiol-PEG-Thiol (SH-PEG-SH) is a high-purity, homobifunctional polyethylene glycol derivative with thiol groups, designed for efficient conjugation of biomolecules. This product offers excellent water solubility and biocompatibility, making it suitable for various biological applications. Its thiol reactivity allows participation in diverse chemical reactions, enhancing the stability and functionality of proteins in drug delivery and diagnostic assays.
  • High purity and biocompatibility
  • Efficient conjugation of biomolecules
  • Excellent water solubility
  • Thiol groups for versatile chemical reactivity
  • Enhances protein stability and half-life
  • Suitable for drug delivery systems and diagnostics
  • Packed in 5 g for convenience
  • Store at -20°C for optimal longevity
Catalog No. NC2455867 Supplier LAYSAN BIO INC Supplier No. SHPEGSH34005G
LAYSAN BIO INC's Thiol-PEG-Thiol (SH-PEG-SH) is a high-purity, homobifunctional polyethylene glycol derivative with thiol groups, designed for efficient conjugation of biomolecules. This product offers excellent water solubility and biocompatibility, making it suitable for various biological applications. Its thiol reactivity allows participation in diverse chemical reactions, enhancing the stability and functionality of proteins in drug delivery and diagnostic assays.
  • High purity and biocompatibility
  • Efficient conjugation of biomolecules
  • Excellent water solubility
  • Thiol groups for versatile chemical reactivity
  • Enhances protein stability and half-life
  • Suitable for drug delivery systems and diagnostics
  • Packed in 5 g for convenience
  • Store at -20°C for optimal longevity