Learn More
Medchemexpress LLC Thiol-PEG1-acid | 1260092-48-5 | 95.0% | 150.20 g·mol⁻¹ | C5H10O3S | 25 MG
SDP
Catalog No. 5000281578
Encompass_Preferred
Thiol-PEG1-acid is a short-chain polyethylene glycol derivative with a terminal thiol and carboxylic acid. It functions as a bifunctional linker and short PEG spacer for bioconjugation, surface modification, and click-chemistry applications, providing a reactive thiol for coupling and a carboxyl group for further derivatization. It is supplied as a laboratory reagent with specified storage recommendations.
  • Contains reactive thiol (-SH) and carboxyl (-COOH) functional groups.
  • Provides a short PEG spacer to minimize steric hindrance.
  • Good solubility in DMSO for common bioconjugation workflows.
  • Suitable for bioconjugation, surface functionalization, and assay reagent development.
  • Stable when stored under recommended conditions noted in the safety data sheet.
Catalog No. 50-002-81578 Supplier Medchemexpress LLC Supplier No. HYW19087725MG
Add to Cart
Add to Cart
Thiol-PEG1-acid is a short-chain polyethylene glycol derivative with a terminal thiol and carboxylic acid. It functions as a bifunctional linker and short PEG spacer for bioconjugation, surface modification, and click-chemistry applications, providing a reactive thiol for coupling and a carboxyl group for further derivatization. It is supplied as a laboratory reagent with specified storage recommendations.
  • Contains reactive thiol (-SH) and carboxyl (-COOH) functional groups.
  • Provides a short PEG spacer to minimize steric hindrance.
  • Good solubility in DMSO for common bioconjugation workflows.
  • Suitable for bioconjugation, surface functionalization, and assay reagent development.
  • Stable when stored under recommended conditions noted in the safety data sheet.