Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thiol-PEG1-acid | 1260092-48-5 | 95.0% | 150.20 g/mol | C5H10O3S | 5 MG
SDP

Catalog No. 5000282216
Encompass_Preferred

Thiol-PEG1-acid is a short polyethylene glycol linker bearing a thiol (-SH) and a terminal carboxyl (-COOH). It serves as a bifunctional reagent for chemical biology and bioconjugation applications that require a small, hydrophilic spacer with orthogonal reactive handles.

  • Contains a reactive thiol (-SH) functional group for selective conjugation.
  • Terminal carboxyl group enables amide or ester bond formation with amines or alcohols.
  • Short PEG spacer improves solubility and reduces steric hindrance in conjugates.
  • Typical purity listed as 95.0%.
  • Soluble in DMSO at 100 mg/mL; may require ultrasonic assistance.
  • Stable as a pure material at -20°C for up to 3 years; in solution store at -80°C for extended stability.

Catalog No. 50-002-82216 Supplier Medchemexpress LLC Supplier No. HYW1908775MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Thiol-PEG1-acid is a short polyethylene glycol linker bearing a thiol (-SH) and a terminal carboxyl (-COOH). It serves as a bifunctional reagent for chemical biology and bioconjugation applications that require a small, hydrophilic spacer with orthogonal reactive handles.

  • Contains a reactive thiol (-SH) functional group for selective conjugation.
  • Terminal carboxyl group enables amide or ester bond formation with amines or alcohols.
  • Short PEG spacer improves solubility and reduces steric hindrance in conjugates.
  • Typical purity listed as 95.0%.
  • Soluble in DMSO at 100 mg/mL; may require ultrasonic assistance.
  • Stable as a pure material at -20°C for up to 3 years; in solution store at -80°C for extended stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.