Learn More
Medchemexpress LLC Thiol-PEG2-acid | 1379649-73-6 | 96.0% | 194.25 | 100 MG
SDP
Catalog No. 5000351022
Encompass_Preferred
Thiol-PEG2-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs themselves contain two different ligands connected by a linker; one ligand is for an E3 ubiquitin ligase and the other is for the target protein, and they exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. For research use only. We do not sell to patients.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-51022 Supplier Medchemexpress LLC Supplier No. HY138524100MG
Thiol-PEG2-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs themselves contain two different ligands connected by a linker; one ligand is for an E3 ubiquitin ligase and the other is for the target protein, and they exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. For research use only. We do not sell to patients.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins