Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thiol-PEG2-acid | 1379649-73-6 | 96.0% | 194.25 | 250 MG
SDP

Catalog No. 5000352284
Encompass_Preferred

Thiol-PEG2-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. It is for research use only.

  • Appearance is liquid
  • Color is colorless to light yellow
  • Soluble in DMSO
  • Shipped at room temperature
  • Pure form can be stored at -20°C for 3 years, 4°C for 2 years
  • In solvent, can be stored at -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-52284 Supplier Medchemexpress LLC Supplier No. 250MGHY138524
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thiol-PEG2-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. It is for research use only.

  • Appearance is liquid
  • Color is colorless to light yellow
  • Soluble in DMSO
  • Shipped at room temperature
  • Pure form can be stored at -20°C for 3 years, 4°C for 2 years
  • In solvent, can be stored at -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.