Learn More
Medchemexpress LLC Thiol-PEG2-acid | 1379649-73-6 | 96.0% | 194.25 | 50 MG
SDP
Catalog No. 5000349472
Encompass_Preferred
Thiol-PEG2-acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These innovative molecules exploit the intracellular ubiquitin-proteasome system for selective degradation of target proteins. Comprising two distinct ligands connected by a linker, PROTACs offer a promising approach for targeted therapeutic development by actively removing disease-causing proteins. Thiol-PEG2-acid serves as a crucial linker component in assembling these complex PROTAC structures.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Facilitates targeted protein degradation via the ubiquitin-proteasome system
Catalog No. 50-003-49472 Supplier Medchemexpress LLC Supplier No. HY13852450MG
Thiol-PEG2-acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These innovative molecules exploit the intracellular ubiquitin-proteasome system for selective degradation of target proteins. Comprising two distinct ligands connected by a linker, PROTACs offer a promising approach for targeted therapeutic development by actively removing disease-causing proteins. Thiol-PEG2-acid serves as a crucial linker component in assembling these complex PROTAC structures.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Facilitates targeted protein degradation via the ubiquitin-proteasome system