Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thiol-PEG2-t-butyl ester | 1398044-50-2 | MFCD22574781 | 96.0% | 250.35 g·mol-1 | C11H22O4S | 5 MG
SDP

Catalog No. 5000262146
Encompass_Preferred

Thiol-PEG2-t-butyl ester is a short polyethylene glycol (PEG2) linker bearing a tert-butyl-protected thiol, designed for use in the synthesis of proteolysis-targeting chimeras (PROTACs). It provides a hydrophilic spacer and a masked thiol functionality for selective deprotection and conjugation in medicinal chemistry workflows.

  • Provides a short PEG2 spacer to improve solubility and flexibility.
  • Contains a tert-butyl-protected thiol for masked thiol chemistry.
  • Suitable for incorporation into bifunctional PROTAC molecules.
  • Available in research-scale pack sizes for synthetic work.
  • Molecular weight ~250.35 g·mol-1 and formula C11H22O4S.

Catalog No. 50-002-62146 Supplier Medchemexpress LLC Supplier No. HY1321005MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thiol-PEG2-t-butyl ester is a short polyethylene glycol (PEG2) linker bearing a tert-butyl-protected thiol, designed for use in the synthesis of proteolysis-targeting chimeras (PROTACs). It provides a hydrophilic spacer and a masked thiol functionality for selective deprotection and conjugation in medicinal chemistry workflows.

  • Provides a short PEG2 spacer to improve solubility and flexibility.
  • Contains a tert-butyl-protected thiol for masked thiol chemistry.
  • Suitable for incorporation into bifunctional PROTAC molecules.
  • Available in research-scale pack sizes for synthetic work.
  • Molecular weight ~250.35 g·mol-1 and formula C11H22O4S.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.