Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thiol-PEG3-acid | 1347750-82-6 | ≥95.0% | 238.30 | 250 MG
SDP

Catalog No. 5000352289
Encompass_Preferred

Thiol-PEG3-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker: one for an E3 ubiquitin ligase and one for the target protein. They exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • For research use only
  • Purity of ≥95.0%
  • Molecular weight of 238.30
  • CAS number 1347750-82-6

Catalog No. 50-003-52289 Supplier Medchemexpress LLC Supplier No. 250MGHY140018
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Thiol-PEG3-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker: one for an E3 ubiquitin ligase and one for the target protein. They exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • For research use only
  • Purity of ≥95.0%
  • Molecular weight of 238.30
  • CAS number 1347750-82-6

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.