Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thiol-PEG4-acid | 749247-06-1 | 95.0% | 25 MG
SDP

Catalog No. 5000338797
Encompass_Preferred

Thiol-PEG4-acid is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. These compounds consist of two distinct ligands connected by a linker: one targeting an E3 ubiquitin ligase and the other targeting a protein for degradation. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • Purity of 95.0%
  • Molecular weight: 282.35
  • Chemical formula: C11H22O6S
  • Appearance: colorless to light yellow liquid
  • Storage at 4°C under nitrogen
  • In solvent, store at -80°C for 6 months or -20°C for 1 month under nitrogen
  • Density: 1.156±0.06 g/cm3

Catalog No. 50-003-38797 Supplier Medchemexpress LLC Supplier No. 25MGHY130213
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thiol-PEG4-acid is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. These compounds consist of two distinct ligands connected by a linker: one targeting an E3 ubiquitin ligase and the other targeting a protein for degradation. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • Purity of 95.0%
  • Molecular weight: 282.35
  • Chemical formula: C11H22O6S
  • Appearance: colorless to light yellow liquid
  • Storage at 4°C under nitrogen
  • In solvent, store at -80°C for 6 months or -20°C for 1 month under nitrogen
  • Density: 1.156±0.06 g/cm3

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.