Learn More
Medchemexpress LLC Thiol-PEG4-NH2 hydrochloride | 2762393-21-3 | 95.0% | 289.82 | 100 MG
SDP
Catalog No. 5000348370
Encompass_Preferred
Thiol-PEG4-NH2 hydrochloride is a PEG-based PROTAC linker suitable for the synthesis of PROTACs. PROTACs are compounds designed to exploit the intracellular ubiquitin-proteasome system, selectively degrading target proteins by connecting a ligand for an E3 ubiquitin ligase and a ligand for the target protein.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-48370 Supplier Medchemexpress LLC Supplier No. HY138446100MG
Thiol-PEG4-NH2 hydrochloride is a PEG-based PROTAC linker suitable for the synthesis of PROTACs. PROTACs are compounds designed to exploit the intracellular ubiquitin-proteasome system, selectively degrading target proteins by connecting a ligand for an E3 ubiquitin ligase and a ligand for the target protein.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins