Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thiol-PEG5-amine hydrochloride | 1446256-16-1 | MFCD29052169 | 98.0% | 333.87 g/mol | C12H28ClNO5S | 5 MG
SDP

Catalog No. 5000262939
Encompass_Preferred

HS-PEG5-CH2CH2NH2 hydrochloride is a PEG-based PROTAC linker supplied as the hydrochloride salt. It features a terminal thiol and a terminal primary amine, enabling orthogonal conjugation strategies for assembly of bifunctional molecules and bioconjugates. The reagent is provided in small laboratory quantities for medicinal chemistry and chemical biology research.

  • Provides thiol and amine functional groups for bioconjugation.
  • PEG5 spacer enhances solubility and flexibility in aqueous and organic media.
  • Hydrochloride salt form improves stability and handling.
  • Suitable for PROTAC linker synthesis and other bifunctional molecule assembly.
  • Manufacturer-stated purity: 98.0%.
  • Molecular weight 333.87 g/mol; molecular formula C12H28ClNO5S.
  • Soluble in DMSO for use in small-scale reactions.

Catalog No. 50-002-62939 Supplier Medchemexpress LLC Supplier No. HY130872A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HS-PEG5-CH2CH2NH2 hydrochloride is a PEG-based PROTAC linker supplied as the hydrochloride salt. It features a terminal thiol and a terminal primary amine, enabling orthogonal conjugation strategies for assembly of bifunctional molecules and bioconjugates. The reagent is provided in small laboratory quantities for medicinal chemistry and chemical biology research.

  • Provides thiol and amine functional groups for bioconjugation.
  • PEG5 spacer enhances solubility and flexibility in aqueous and organic media.
  • Hydrochloride salt form improves stability and handling.
  • Suitable for PROTAC linker synthesis and other bifunctional molecule assembly.
  • Manufacturer-stated purity: 98.0%.
  • Molecular weight 333.87 g/mol; molecular formula C12H28ClNO5S.
  • Soluble in DMSO for use in small-scale reactions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.