Learn More
Medchemexpress LLC Thiol-PEG6-alcohol | 194425-46-2 | ≥97.0% | 1 G
SDP
Catalog No. 5000338178
Encompass_Preferred
Thiol-PEG6-alcohol is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs function by connecting two distinct ligands, one for an E3 ubiquitin ligase and the other for a target protein, enabling the selective degradation of target proteins via the intracellular ubiquitin-proteasome system. This product is intended for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits ubiquitin-proteasome system for selective protein degradation
Catalog No. 50-003-38178 Supplier Medchemexpress LLC Supplier No. HY1305801G
Thiol-PEG6-alcohol is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs function by connecting two distinct ligands, one for an E3 ubiquitin ligase and the other for a target protein, enabling the selective degradation of target proteins via the intracellular ubiquitin-proteasome system. This product is intended for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits ubiquitin-proteasome system for selective protein degradation