Learn More
Medchemexpress LLC Thiol-PEG6-alcohol | 194425-46-2 | ≥97.0% | 250 MG
SDP
Catalog No. 5000343017
Encompass_Preferred
Thiol-PEG6-alcohol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker.
  • Used in the synthesis of PROTACs.
Catalog No. 50-003-43017 Supplier Medchemexpress LLC Supplier No. 250MGHY130580
Add to Cart
Add to Cart
Thiol-PEG6-alcohol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker.
  • Used in the synthesis of PROTACs.