Learn More
Medchemexpress LLC Thiol-PEG6-alcohol | 194425-46-2 | ≥97.0% | 50 MG
SDP
Catalog No. 5000338837
Encompass_Preferred
Thiol-PEG6-alcohol is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are molecules containing two different ligands connected by a linker, where one ligand targets an E3 ubiquitin ligase and the other targets a specific protein. They function by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Leverages the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-38837 Supplier Medchemexpress LLC Supplier No. HY13058050MG
Thiol-PEG6-alcohol is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are molecules containing two different ligands connected by a linker, where one ligand targets an E3 ubiquitin ligase and the other targets a specific protein. They function by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Leverages the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins