Learn More
Medchemexpress LLC Thiol-PEG6-alcohol (HS-PEG6-OH) | 194425-46-2 | ≥97.0% | 100 MG
SDP
Catalog No. 5000338484
Encompass_Preferred
Thiol-PEG6-alcohol is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs feature two distinct ligands connected by a linker, one targeting an E3 ubiquitin ligase and the other binding to the target protein. This mechanism allows PROTACs to leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • PEG-based PROTAC linker.
  • Used in the synthesis of PROTACs.
  • PROTACs utilize the ubiquitin-proteasome system.
  • Designed for selective degradation of target proteins.
Catalog No. 50-003-38484 Supplier Medchemexpress LLC Supplier No. HY130580100MG
Thiol-PEG6-alcohol is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs feature two distinct ligands connected by a linker, one targeting an E3 ubiquitin ligase and the other binding to the target protein. This mechanism allows PROTACs to leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • PEG-based PROTAC linker.
  • Used in the synthesis of PROTACs.
  • PROTACs utilize the ubiquitin-proteasome system.
  • Designed for selective degradation of target proteins.