Learn More
Medchemexpress LLC Thiol-PEG8-acid | 866889-02-3 | 97.0% | 250 MG
SDP
Catalog No. 5000341457
Encompass_Preferred
Thiol-PEG8-acid is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs function by connecting two different ligands, one for an E3 ubiquitin ligase and another for the target protein. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-41457 Supplier Medchemexpress LLC Supplier No. 250MGHY130542
Thiol-PEG8-acid is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs function by connecting two different ligands, one for an E3 ubiquitin ligase and another for the target protein. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins