Learn More
Biotium Thiol Reactive Dyes Reactive CF Dyes Other Dyes Biotinylation Reagents Reactive CF Dyes CF405 1/EA
Catalog No. 5000501587
Encompass_Preferred
CF Dye Maleimides are thiol-reactive fluorescent dyes Maleimides are commonly used to label proteins on free thiol groups CF dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes Blue fluorescent CF405S dye has excitation/emission at 404/431 nm
Catalog No. 50-005-01587 Supplier Biotium Supplier No. 92030
CF Dye Maleimides are thiol-reactive fluorescent dyes Maleimides are commonly used to label proteins on free thiol groups CF dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes Blue fluorescent CF405S dye has excitation/emission at 404/431 nm