Learn More
Biotium Thiol Reactive Dyes Reactive CF Dyes Other Dyes Biotinylation Reagents Reactive CF Dyes CF583 1/EA
Catalog No. 5000501551
Encompass_Preferred
CF Dye Maleimides are thiol-reactive fluorescent dyes Maleimides are commonly used to label proteins on free thiol groups CF Dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes Red fluorescent CF583R dye has excitation/emission at 585/609 nm
Catalog No. 50-005-01551 Supplier Biotium Supplier No. 96107
CF Dye Maleimides are thiol-reactive fluorescent dyes Maleimides are commonly used to label proteins on free thiol groups CF Dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes Red fluorescent CF583R dye has excitation/emission at 585/609 nm