Learn More
Enzo Life Sciences Thiolutin (5mg). CAS: 87-11-6
SDP
Supplier:  Enzo Life Sciences BMLCT1070005
Encompass_Preferred
Sulfur containing antibiotic. Potent inhibitor of bacterial and yeast RNA polymerases. Inhibits mRNA chain elongation. Inhibits adhesion of endothelial cells (HUVECs) to vitronectin (IC50=0.83 µM) thereby inhibiting tumor cell-induced angiogenesis in vivo. Inhibits transcription in C. albicans. Inhibits microbiological growth in beer. Active against soil borne pathogens. Alternative name: 6-(Acetamido)-4-methyl-1 2-dithiolo 4 3-b -pyrrol-5(4H)-one. Purity: ≥95% (HPLC, TLC). Solubility: Soluble in DMSO (5mg/ml), very slightly soluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-0482