Learn More
Medchemexpress LLC Thiolutin (acetopyrrothin) | 87-11-6 | 1 ML
SDP
Catalog No. 5000419432
Encompass_Preferred
Thiolutin (Acetopyrrothin) is a sulfur-containing antibiotic and a potent inhibitor of bacterial and yeast RNA polymerases. It is produced by Streptomyces. Thiolutin inhibits AMSH (IC50 = 4 μM) and Rpn11 (IC50 = 0.53 μM). It acts as a dual inhibitor of BRCC36 and the NLRP3 inflammasome, demonstrating anti-inflammatory effects and effectively suppressing the interaction between BRCC36 and HMGCR, thereby inhibiting HCC growth.
  • Potent inhibitor of bacterial and yeast RNA polymerases.
  • Inhibits AMSH (IC50 = 4 μM) and Rpn11 (IC50 = 0.53 μM).
  • Dual inhibitor of BRCC36 and the NLRP3 inflammasome, exhibiting anti-inflammatory effects.
  • Suppresses the interaction between BRCC36 and HMGCR, inhibiting HCC growth.
  • Attenuates pyroptosis and NLRP3 inflammasome activation.
  • Alleviates renal injury and inflammatory process in IgAN.
  • Anti-angiogenic compound that can ease doxorubicin-induced cardiotoxicity (DOXIC).
Catalog No. 50-004-19432 Supplier Medchemexpress LLC Supplier No. HYN671210MM/1ML
Thiolutin (Acetopyrrothin) is a sulfur-containing antibiotic and a potent inhibitor of bacterial and yeast RNA polymerases. It is produced by Streptomyces. Thiolutin inhibits AMSH (IC50 = 4 μM) and Rpn11 (IC50 = 0.53 μM). It acts as a dual inhibitor of BRCC36 and the NLRP3 inflammasome, demonstrating anti-inflammatory effects and effectively suppressing the interaction between BRCC36 and HMGCR, thereby inhibiting HCC growth.
  • Potent inhibitor of bacterial and yeast RNA polymerases.
  • Inhibits AMSH (IC50 = 4 μM) and Rpn11 (IC50 = 0.53 μM).
  • Dual inhibitor of BRCC36 and the NLRP3 inflammasome, exhibiting anti-inflammatory effects.
  • Suppresses the interaction between BRCC36 and HMGCR, inhibiting HCC growth.
  • Attenuates pyroptosis and NLRP3 inflammasome activation.
  • Alleviates renal injury and inflammatory process in IgAN.
  • Anti-angiogenic compound that can ease doxorubicin-induced cardiotoxicity (DOXIC).