Learn More
Medchemexpress LLC Thiolutin (Acetopyrrothin) | 87-11-6 | 10 MG
SDP
Catalog No. 5000418003
Encompass_Preferred
Thiolutin (Acetopyrrothin) is a sulfur-containing antibiotic and a potent inhibitor of bacterial and yeast RNA polymerases. Produced by Streptomyces, it functions as a dual inhibitor of BRCC36 and the NLRP3 inflammasome, demonstrating anti-inflammatory and anti-angiogenic properties.
  • Sulfur-containing antibiotic
  • Potent inhibitor of bacterial and yeast RNA polymerases
  • Inhibits AMSH and Rpn11
  • Dual inhibitor of BRCC36 and the NLRP3 inflammasome
  • Anti-inflammatory effects
  • Inhibits HCC growth by suppressing BRCC36 and HMGCR interaction
  • Attenuates pyroptosis and NLRP3 inflammasome activation
  • Alleviates renal injury and inflammatory processes in IgAN
  • Acts as an anti-angiogenic compound
  • Eases Doxorubicin-induced cardiotoxicity
Catalog No. 50-004-18003 Supplier Medchemexpress LLC Supplier No. HYN671210MG
Thiolutin (Acetopyrrothin) is a sulfur-containing antibiotic and a potent inhibitor of bacterial and yeast RNA polymerases. Produced by Streptomyces, it functions as a dual inhibitor of BRCC36 and the NLRP3 inflammasome, demonstrating anti-inflammatory and anti-angiogenic properties.
  • Sulfur-containing antibiotic
  • Potent inhibitor of bacterial and yeast RNA polymerases
  • Inhibits AMSH and Rpn11
  • Dual inhibitor of BRCC36 and the NLRP3 inflammasome
  • Anti-inflammatory effects
  • Inhibits HCC growth by suppressing BRCC36 and HMGCR interaction
  • Attenuates pyroptosis and NLRP3 inflammasome activation
  • Alleviates renal injury and inflammatory processes in IgAN
  • Acts as an anti-angiogenic compound
  • Eases Doxorubicin-induced cardiotoxicity