Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiolutin, Tocris Bioscience™
SDP

Catalog No. 15671
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-671 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 15-671 Supplier R&D Systems Supplier No. 1567/1

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Bacterial RNA polymerase inhibitor

Antibiotic; inhibits bacterial RNA polymerase. Inhibits adhesion of HUVEC cells to vitronectin (IC50 = 0.83 mM) and subsequently reduces paxillin levels. Suppresses tumor cell-induced angiogenesis.

Chemical Identifiers

CAS 87-11-6
Molecular Formula C8H8N2O2S2
Molecular Weight (g/mol) 228.284
InChI Key MHMRAFONCSQAIA-UHFFFAOYSA-N
Synonym thiolutin, acetopyrrothin, acetopyrrothine, n-4,5-dihydro-4-methyl-5-oxo-1,2-dithiolo 4,3-b pyrrol-6-yl acetamide, 3-acetamido-5-methylpyrrolin-4-one 4,3-d-1,2-dithiole, 6-acetamido-4-methyl-1,2-dithiolo 4,3-b pyrrol-5 4h-one, 6-acetylamino-4-methyl-1,2-dithiolo 4,3-b pyrrol-5 4h-one, 1,2-dithiolo 4.3-b pyrrol-5 4h-one, 6-acetamido-4-methyl
PubChem CID 6870
IUPAC Name N-(4-methyl-5-oxodithiolo[4,3-b]pyrrol-6-yl)acetamide
SMILES CC(=O)NC1=C2C(=CSS2)N(C1=O)C

Specifications

Quantity 1 mg
Formula Weight 228.28
Percent Purity >98%
Chemical Name or Material Thiolutin
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.