Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiomorpholinoacetic Acid 1',1'-Dioxide Monohydrate 98.0+%, TCI America™
SDP

Catalog No. T20505G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T20505G 5 g
T205025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T20505G Supplier TCI America Supplier No. T20505G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 155480-08-3
Molecular Formula C6H11NO4S
Molecular Weight (g/mol) 193.217
MDL Number MFCD00121261
InChI Key CEABBIBOUBCOPV-UHFFFAOYSA-N
Synonym 4-(Carboxymethyl)thiomorpholine 1,1-Dioxide, (1,1-Dioxothiomorpholino)acetic Acid
PubChem CID 2806236
IUPAC Name 2-(1,1-dioxo-1,4-thiazinan-4-yl)acetic acid
SMILES C1CS(=O)(=O)CCN1CC(=O)O

Specifications

Melting Point 177°C
Color White
Quantity 5 g
Formula Weight 193.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Thiomorpholinoacetic Acid 1′,1′-Dioxide Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.