Learn More
Cayman Chemical Thiomuscimol, Purity: ≥95%, CAS: 62020-54-6, 10 mg
SDP
Catalog No. 502983400
Encompass_Preferred
A GABAA receptor agonist (IC50 = 19 nM) that has been used as a photoaffinity label for the purification and identification of GABA binding sites within the GABAA receptor complex
Catalog No. 50-298-3400 Supplier Cayman Chemical Supplier No. 1885710 MG
A GABAA receptor agonist (IC50 = 19 nM) that has been used as a photoaffinity label for the purification and identification of GABA binding sites within the GABAA receptor complex