Learn More
Selleck Chemical LLC Thiomyristoyl 2mg 1429749-41-6 TM
SDP
Catalog No. NC1300793
Encompass_Preferred
Thiomyristoyl (TM) is a potent and specific SIRT2 inhibitor with an IC50 of 28 nM. It inhibits SIRT1 with an IC50 value of 98 ?M and does not inhibit SIRT3 even at 200 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1300793 Supplier Selleck Chemical LLC Supplier No. S82452MG
Thiomyristoyl (TM) is a potent and specific SIRT2 inhibitor with an IC50 of 28 nM. It inhibits SIRT1 with an IC50 value of 98 ?M and does not inhibit SIRT3 even at 200 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.