Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thionicotinamide, 98%

Catalog No. p-7081244
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1114414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1114414 Supplier Thermo Scientific Chemicals Supplier No. A1114414

CAS: 4621-66-3 | C6H6N2S | 138.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4621-66-3
Molecular Formula C6H6N2S
Molecular Weight (g/mol) 138.19
MDL Number MFCD00006399
InChI Key XQWBMZWDJAZPPX-UHFFFAOYSA-N
Synonym thionicotinamide, 3-pyridinecarbothioamide, nicotinothioamide, 3-thiocarbamoylpyridine, 3-thioamidopyridine, nicotinamide, thio, 3-pyridylthioformamide, 3-pyridinethiocarboxamide, thio-3-pyridinecarboxamide, unii-6a0hxx0753
PubChem CID 737155
IUPAC Name pyridine-3-carbothioamide
SMILES NC(=S)C1=CC=CN=C1

Specifications

Melting Point ∼190°C (decomposition)
Flash Point 160°C (320°F)
Quantity 25 g
Beilstein 109593
Formula Weight 138.19
Percent Purity 98%
Chemical Name or Material Thionicotinamide
Hazard Category Warning
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 225-036-6
RTECSNumber QS4488000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.